Extran® rengøringsmidler, flydende koncentrater, til manual brug (MA)
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.07553.2500 1.07553.9010 1.07553.9025 1.07555.2500 1.07555.9010 1.07555.9025 1.07555.9200 1.07555.5000 1.07555.1000 1.40000.2500 1.40000.9010 1.40000.9025 1.40000.5000 1.07553.5000
1.07553.2500EA 585.98 DKK
1.07553.2500 1.07553.9010 1.07553.9025 1.07555.2500 1.07555.9010 1.07555.9025 1.07555.9200 1.07555.5000 1.07555.1000 1.40000.2500 1.40000.9010 1.40000.9025 1.40000.5000 1.07553.5000
Extran® rengøringsmidler, flydende koncentrater, til manual brug (MA)
Renseopløsninger Detergenter og Overflade Rengøringsmidler
A range of cleansers, for manual cleaning, that have been developed to meet the cleaning requirements for glassware and small equipment in the laboratory.
  • Prevents carry-over of residues to the next analysis
  • Chlorine-free
These manual cleansers are supplied as concentrates and the cleaning solutions are simply made up with tap water. Typically immersion for up to 2 hours is sufficient, but for particularly difficult residues, the time period can be extended. Alternatively, warming the solution or use with an ultrasonic bath can speed up the process.More Product Information