Pyrogallol red indicator for metal titration

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3',4',5',6'-Tetrahydroxyspiro[benzo[c][1,2]oxathiole-3,9'-xanthene] 1,1-dioxide

M-Clarity™ quality level = MQ200

1.07534.0001
1.07534.0001EA 1500 DKK
1.07534.0001
Pyrogallol red indicator for metal titration
Pyrogallol red
Formel: C₁₉H₁₂O₈S
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005046
CAS-nummer: 32638-88-3

Bestil nu

Specifikationer Testresultater

Appearance of solution conforms
Loss on drying (110 °C) ≤ 5.0 %
Suitability as indicator (for metal titration) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR