Natriumperoxodisulfat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt

M-Clarity™ quality level = MQ300

1.06609.0500 1.06609.1000 1.06609.9025 1.06609.5000 1.06609.9012
1.06609.0500EA 358 DKK
1.06609.0500 1.06609.1000 1.06609.9025 1.06609.5000 1.06609.9012
Natriumperoxodisulfat, EMSURE® for analysis, Supelco®
Sodium peroxodisulphate
Formel: Na₂S₂O₈
Smeltepunkt: 100 °C
Massefylde: 2,4 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003501
CAS-nummer: 7775-27-1
EINECS: 231-892-1
UN nummer: 1505
ADR: 5.1,III
Merck Index: 13,08729

Bestil nu

Specifikationer Testresultater

Assay (iodometric) ≥ 99.0 %
Identity passes test
Chloride (Cl) ≤ 0.01 %
Heavy metals (as Pb) ≤ 0.003 %
NH₄ (Ammonium) ≤ 0.05 %
Cr (Chromium) ≤ 0.001 %
Cu (Copper) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
Mn (Manganese) ≤ 0.0001 %
Pb (Lead) ≤ 0.001 %
Loss on drying (105 °C; 3 hours) ≤ 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR