Natriumfluorid, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ100

1.06450.0025
1.06450.0025EA 1710 DKK
1.06450.0025
Natriumfluorid, Suprapur® for trace analysis, Supelco®
Sodium fluoride
Formel: NaF
Kogepunkt: 1700 °C (1013 hPa)
Smeltepunkt: 993 °C
Massefylde: 2,8 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003524
CAS-nummer: 7681-49-4
EINECS: 231-667-8
UN nummer: 1690
ADR: 6.1,III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 42 g/l (20 °C)
Melting Point 993 °C
Molar Mass 41.98 g/mol
Bulk Density 800 kg/m3
Boiling Point 1704 °C
Vapor Pressure 1 hPa (1077 °C)
Density 2.8 g/cm3 (20 °C)
pH 10.2 (40 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR