Natriumcarbonat, anhydrous, Suprapur® for trace analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Calcined soda, Carbonic acid disodium salt

M-Clarity™ quality level = MQ100

1.06395.0050 1.06395.0500
1.06395.0050EA 1480 DKK
1.06395.0050 1.06395.0500
Natriumcarbonat, anhydrous, Suprapur® for trace analysis, Supelco®
Sodium carbonate
Formel: Na₂CO₃
Kogepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 854 °C
Massefylde: 2,53 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003494
CAS-nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 220 g/l (20 °C)
Melting Point 854 °C
Molar Mass 105.99 g/mol
Bulk Density 1100 kg/m3
Boiling Point 1600 °C (decomposition)
Density 2.53 g/cm3 (20 °C)
pH 11.5 (50 g/l, H2O, 25 °C)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR