Natriumhydrogensulfat monohydrat, EMSURE® for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Sodium bisulphate monohydrate

M-Clarity™ quality level = MQ300

1.06352.0500 1.06352.9050
1.06352.0500EA 558 DKK
1.06352.0500 1.06352.9050
Natriumhydrogensulfat monohydrat, EMSURE® for analysis, Supelco®
Natriumhydrogensulfat monohydrat
Formel: NaHSO₄·H₂O
Smeltepunkt: 183 °C
Massefylde: 2,1 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149210
CAS-nummer: 10034-88-5
EINECS: 231-665-7
UN nummer: 3260
ADR: 8,III
Merck Index: 13,08658

Bestil nu

Specifikationer Testresultater

Solubility 1080 g/l (20 °C)
Melting Point 183 °C (decomposition)
Molar Mass 138.07 g/mol
Bulk Density 900 - 970 kg/m3
Density 2.12 g/cm3 (20 °C)
pH 1 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR