MOPS, Millipore®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid

M-Clarity™ quality level = MQ100

1.06129.0025 1.06129.0250 1.06129.9010
1.06129.0025EA 376 DKK
1.06129.0025 1.06129.0250 1.06129.9010
MOPS, Millipore®
MOPS
Formel: C₇H₁₅NO₄S
Molekylvægt: 209.27 g/mol
Smeltepunkt: > 250 °C
Flammepunkt: 110 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006183
CAS-nummer: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR