Methylethylketon, EMPLURA®, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethylmethylketon, Butanon

M-Clarity™ quality level = MQ200

1.06014.2500 1.06014.6025 1.06014.9190
1.06014.2500EA 852 DKK
1.06014.2500 1.06014.6025 1.06014.9190
Methylethylketon, EMPLURA®, Supelco®
Methylethylketon
Formel: C₂H₅COCH₃
Kogepunkt: 79,6 °C (1013 hPa)
Smeltepunkt: –86 °C
Massefylde: 0,8054 g/cm³ (20 °C)
Flammepunkt: –4 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011648
CAS-nummer: 78-93-3
EINECS: 201-159-0
UN nummer: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR