Kaolin, powder, EMPROVE® EXPERT BP, Ph. Eur., extra_pure excipient_suitable, Use_internal Bacterial_low, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: China Clay, Hydrated aluminum silicate, Clay

1.05006.9025
1.05006.9025EA 0 DKK
1.05006.9025
Kaolin, powder, EMPROVE® EXPERT BP, Ph. Eur., extra_pure excipient_suitable, Use_internal Bacterial_low, SAFC®
Kaolin
Formel: Al₂SiO₇·2H₂O
Molekylvægt: 258.16 g/mol
Smeltepunkt: 1760
Massefylde: 0.5 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00062311
CAS-nummer: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR