Kaliumdichromat, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ300

1.04864.0500 1.04864.1000
1.04864.0500EA 1670 DKK
1.04864.0500 1.04864.1000
Kaliumdichromat, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Potassium dichromate

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues.

Formel: K₂Cr₂O₇
Kogepunkt: 500 °C (1013 hPa)
Smeltepunkt: 398 °C
Massefylde: 2,67 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011367
CAS-nummer: 7778-50-9
EINECS: 231-906-6
UN nummer: 3086
ADR: 6.1 (5.1),II
REACH: 01-2119454792-32
Merck Index: 12,07790

Bestil nu

Specifikationer Testresultater

Solubility 130 g/l (20 °C)
Melting Point 398 °C
Molar Mass 294.19 g/mol
Bulk Density 1250 kg/m3
Boiling Point 500 °C (1013 hPa)
Density 2.69 g/cm3 (20 °C)
pH 3.6 (100 g/l, H2O)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR