indicator for metal titration, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalen-3-carboxylsyre dihydrat

M-Clarity™ quality level = MQ200

1.04595.0005 1.04595.0025
1.04595.0005EA 1770 DKK
1.04595.0005 1.04595.0025
indicator for metal titration, Supelco®
Patton-reeders reagent (Calconcarboxylic acid dihydrate)
Formel: C₂₁H₁₄N₂O₇S·2H₂O
Molekylvægt: 474.45 g/mol
MDL Number: MFCD00004078
CAS-nummer: 3737-95-9
EINECS: 223-117-0

Bestil nu

Specifikationer Testresultater

Appearance of solution passes test
Water (according to Karl Fischer) ≤ 9%
Suitability as indicator (for metal titration) passes test
Bulk Density 400 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR