Kieselgel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M-Clarity™ quality level = MQ200
1.03804.0001
1.03804.0001EA 1950 DKK
1.03804.0001
Kieselgel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Silica gel
Emballering: 50 sachets in a metal can
Formel: SiO₂
Smeltepunkt: > 1000 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 63231-67-4
Bestil nu
Specifikationer Testresultater
Water absorption capacity (24 h, 80 % relative humidity) ≥ 20 %
Loss on drying (140°C, 3 h) ≤ 10 %
Water Absorption 250 g/kg (20 °C)