4-Amino-N,N-dimethylanilin 2 HCl analytical reagent, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

M-Clarity™ quality level = MQ100

1.03067.0025
1.03067.0025EA 862 DKK
1.03067.0025
4-Amino-N,N-dimethylanilin 2 HCl analytical reagent, Supelco®
4-Amino-N,N-dimethylanilin 2 HCl
Formel: (CH₃)₂NC₆H₄NH₂·2HCl
Smeltepunkt: 208…212 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012991
CAS-nummer: 536-46-9
EINECS: 208-635-7
UN nummer: 2811
ADR: 6.1,II
Merck Index: 13,03280

Bestil nu

Specifikationer Testresultater

Solubility 2000 g/l (20 °C)
Melting Point 208 - 212 °C (decomposition)
Molar Mass 209.11 g/mol
Bulk Density 500 kg/m3
pH 1.6 (50 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR