Kobber(II)oxid, wire ACS (0.65x6 mm) elementary_analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Kobberoxid

MQ100

1.02767.0250
1.02767.0250EA 1340 DKK
1.02767.0250
Kobber(II)oxid, wire ACS (0.65x6 mm) elementary_analysis, Supelco®
Kobber(II)oxid

Surface: CuO
Core: Cu₂O

Formel: CuO
Molekylvægt: 79.55 g/mol
Kogepunkt: 2000
Smeltepunkt: 1326
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010979
CAS-nummer: 1317-38-0
EINECS: 215-269-1
UN nummer: 3077
ADR: 9 III
Merck Index: 13,02674

Bestil nu

Specifikationer Testresultater

Assay (oxygen demand as CuO) ≥ 55.0 %
Chloride (Cl) ≤ 0.005 %
Total carbon (C) ≤ 0.002 %
Total nitrogen (N) ≤ 0.002 %
Total sulfur (as SO₄) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR