Kobber(II)oxid, granular for elementary analysis pH 7 (50 g/l, H₂O, 20 °C) (slurry), Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Kobberoxid

M-Clarity™ quality level = MQ100

1.02764.0250 1.02764.2500
1.02764.0250EA 3050 DKK
1.02764.0250 1.02764.2500
Kobber(II)oxid, granular for elementary analysis pH 7 (50 g/l, H₂O, 20 °C) (slurry), Supelco®
Kobber(II)oxid
Formel: CuO
Kogepunkt: 2000 °C (1013 hPa)
Smeltepunkt: 1326 °C (dec.)
Massefylde: 500 kg/m3
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010979
CAS-nummer: 1317-38-0
EINECS: 215-269-1
UN nummer: 3077
ADR: 9,III
Merck Index: 13,02674

Bestil nu

Specifikationer Testresultater

Assay (complexometric) ≥ 99
Chloride (Cl) ≤ 0.005 %
Total sulfur (as SO₄) ≤ 0.015 %
Total nitrogen (N) ≤ 0.002 %
C (Carbon) ≤ 0.01 %
Bulk density 240 - 270 g/100 ml
Particle size (0.71-2 mm) ≥ 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR