Kobber(I)chlorid, EMSURE® ACS for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Cuprous chloride, Copper chloride

M-Clarity™ quality level = MQ300

1.02739.0250 1.02739.9025
1.02739.0250EA 6630 DKK
1.02739.0250 1.02739.9025
Kobber(I)chlorid, EMSURE® ACS for analysis, Supelco®
Kobber(I)chlorid
Formel: ClCu
Kogepunkt: 1367 °C (1013 hPa)
Smeltepunkt: 430 °C
Massefylde: 4,145 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010971
CAS-nummer: 7758-89-6
EINECS: 231-842-9
UN nummer: 2802
ADR: 8,III
Merck Index: 13,02690

Bestil nu

Specifikationer Testresultater

Solubility 0.001 - 0.1 g/l
Melting Point 422 °C
Molar Mass 99 g/mol
Bulk Density 1600 - 1800 kg/m3
Boiling Point 1367 °C
Density 4.14 g/cm3 (25 °C)
pH 5 (50 g/l, H2O, 20 °C) (slurry)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR