Calciumsulfat dihydrat, EMPROVE® ESSENTIAL Ph. Eur., BP, E516 suitable for use as excipient, precipitated, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Plaster of Paris

1.02160.9050
1.02160.9050EA 0 DKK
1.02160.9050
Calciumsulfat dihydrat, EMPROVE® ESSENTIAL Ph. Eur., BP, E516 suitable for use as excipient, precipitated, SAFC®
Calciumsulfat dihydrat
Formel: CaSO₄·2H₂O
Massefylde: 2,32 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149625
CAS-nummer: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR