di-Ammoniumperoxodisulfat (APS), Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Peroxidisulphuric acid diammonium salt, Ammonium persulphate

M-Clarity™ quality level = MQ200

1.01200.1000 1.01200.5000 1.01200.9025
1.01200.1000EA 652 DKK
1.01200.1000 1.01200.5000 1.01200.9025
di-Ammoniumperoxodisulfat (APS), Supelco®
di-Ammoniumperoxodisulfat (APS)
Formel: (NH₄)₂S₂O₈
Smeltepunkt: 120 °C
Massefylde: 1,98 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003390
CAS-nummer: 7727-54-0
EINECS: 231-786-5
UN nummer: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR