Ammoniumbromid, EMSURE® ACS for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


M-Clarity™ quality level = MQ300

1.01125.1000 1.01125.9025
1.01125.1000EA 1020 DKK
1.01125.1000 1.01125.9025
Ammoniumbromid, EMSURE® ACS for analysis, Supelco®
Ammonium bromide
Formel: H₄BrN
Molekylvægt: 97.94 g/mol
Smeltepunkt: 542
Massefylde: 2.42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011419
CAS-nummer: 12124-97-9
EINECS: 235-183-8
Merck Index: 13,00505

Bestil nu

Specifikationer Testresultater

Solubility 742 g/l (20 °C)
Melting Point 542 °C (closed capillary tube)
Molar Mass 97.94 g/mol
Bulk Density 1100 kg/m3
Vapor Pressure 1.3 hPa (198 °C)
Density 2.43 g/cm3 (20 °C)
pH 4.8 (50 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR