2-Propanol, LiChrosolv®, gradient LC, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopropylalkohol

MQ100

1.01040.1000 1.01040.2500 1.01040.4000 1.01040.9030 1.01040.9185
1.01040.1000EA 366.3 DKK
1.01040.1000 1.01040.2500 1.01040.4000 1.01040.9030 1.01040.9185
2-Propanol, LiChrosolv®, gradient LC, Supelco®
2-Propanol

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formel: (CH₃)₂CHOH
Molekylvægt: 60.1 g/mol
Kogepunkt: 82
Smeltepunkt: –89 °C
Massefylde: 0.786 g/cm³ (20 °C)
Flammepunkt: 12 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011674
CAS-nummer: 67-63-0
EINECS: 200-661-7
UN nummer: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR