Svovlsyre 25% (by alkalimetry), EMSURE® for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ300

1.00716.1000 1.00716.9025
1.00716.1000EA 259 DKK
1.00716.1000 1.00716.9025
Svovlsyre 25% (by alkalimetry), EMSURE® for analysis, Supelco®
Svovlsyre
Formel: H₂SO₄
Kogepunkt: ∼100 °C (1013 hPa)
Massefylde: 1,1…1.4 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
UN nummer: 2796
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Aluminum (Al) 0.05 ppm max.
Ammonium (NH4) 2 ppm max.
Arsenic (As) 0.01 ppm max.
Barium (Ba) 0.05 ppm max.
Beryllium (Be) 0.01 ppm max.
Bismuth (Bi) 0.1 ppm max.
Cadmium (Cd) 0.02 ppm max.
Calcium (Ca) 0.2 ppm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR