ortho-Phosphorsyre 85% ≥85%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phosphorsyre, Fosforsyre

M-Clarity™ quality level = MQ300

1.00573.1000 1.00573.2500 1.00573.9025 1.00573.2510 1.00573.9200 1.00573.0510
1.00573.1000EA 459.33 DKK
1.00573.1000 1.00573.2500 1.00573.9025 1.00573.2510 1.00573.9200 1.00573.0510
ortho-Phosphorsyre 85% ≥85%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
ortho-Phosphorsyre
Formel: H₃PO₄
Smeltepunkt: 28 °C
Massefylde: 1,71…1.87 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011340
CAS-nummer: 7664-38-2
EINECS: 231-633-2
UN nummer: 1805
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Melting Point 21 °C
Boiling Point 158 °C
Vapor Pressure 2 hPa (20 °C)
Density 1.71 g/cm3 (20 °C)
pH  - 0.5 (100 g/l, H2O, 20 °C)
Viscosity kinematic 30.5 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR