Flussyre 48%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Hydrogenfluorid, opløsning

M-Clarity™ quality level = MQ300

1.00334.0500 1.00334.1000 1.00334.5000 1.00334.2500
1.00334.0500EA 516 DKK
1.00334.0500 1.00334.1000 1.00334.5000 1.00334.2500
Flussyre 48%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Flussyre
Formel: HF
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011346
CAS-nummer: 7664-39-3
EINECS: 231-634-8
UN nummer: 1790
ADR: 8 (6.1),II

Bestil nu

Specifikationer Testresultater

Assay (acidimetric) 48.0 - 51.0 %
Color ≤ 10 Hazen
Chloride (Cl) ≤ 1 ppm
Hexafluorosilicate (SiF₆) ≤ 50 ppm
Nitrate (NO₃) ≤ 5 ppm
Phosphate (PO₄) ≤ 0.5 ppm
Sulfate and sulphite (as SO₄) ≤ 5 ppm
Heavy metals (as Pb) ≤ 0.5 ppm
Ag (Silver) ≤ 0.020 ppm
Al (Aluminium) ≤ 0.050 ppm
As (Arsenic) ≤ 0.030 ppm
Au (Gold) ≤ 0.100 ppm
B (Boron) ≤ 0.050 ppm
Ba (Barium) ≤ 0.010 ppm
Be (Beryllium) ≤ 0.020 ppm
Bi (Bismuth) ≤ 0.020 ppm
Ca (Calcium) ≤ 0.200 ppm
Cd (Cadmium) ≤ 0.010 ppm
Co (Cobalt) ≤ 0.020 ppm
Cr (Chromium) ≤ 0.010 ppm
Cu (Copper) ≤ 0.020 ppm
Fe (Iron) ≤ 0.100 ppm
Ga (Gallium) ≤ 0.020 ppm
Ge (Germanium) ≤ 0.020 ppm
In (Indium) ≤ 0.020 ppm
K (Potassium) ≤ 0.100 ppm
Li (Lithium) ≤ 0.010 ppm
Mg (Magnesium) ≤ 0.100 ppm
Mn (Manganese) ≤ 0.030 ppm
Mo (Molybdenum) ≤ 0.020 ppm
Na (Sodium) ≤ 0.200 ppm
Ni (Nickel) ≤ 0.020 ppm
Pb (Lead) ≤ 0.020 ppm
Pt (Platinum) ≤ 0.200 ppm
Sn (Tin) ≤ 0.100 ppm
Sr (Strontium) ≤ 0.020 ppm
Ti (Titanium) ≤ 0.020 ppm
Tl (Thallium) ≤ 0.020 ppm
V (Vanadium) ≤ 0.020 ppm
Zn (Zinc) ≤ 0.050 ppm
Zr (Zirconium) ≤ 0.020 ppm
Residue on ignition (as sulfates) ≤ 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR