Eddikesyre 96% ≥96%, EMSURE® for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

M-Clarity™ quality level = MQ300

1.00062.1011 1.00062.2500 1.00062.2511 1.00062.9025 1.00062.1000
1.00062.1011EA 422 DKK
1.00062.1011 1.00062.2500 1.00062.2511 1.00062.9025 1.00062.1000
Eddikesyre 96% ≥96%, EMSURE® for analysis, Supelco®
Eddikesyre
Formel: H₃CCOOH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
UN nummer: 2789
ADR: 8 (3),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Melting Point 16.6 °C
Boiling Point 117.9 °C (1013 hPa)
Vapor Pressure 15.2 hPa (20 °C)
Flash Point 39 °C
Explosion Limit 4 - 19.9 %(V)
Density 1.05 g/cm3 (20 °C)
pH 2.5 (10 g/l, H2O 20 °C)
Ignition Point 463 °C
Evaporation number 24


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR