Acetonitril, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

M-Clarity™ quality level = MQ300

1.00003.1000 1.00003.2500 1.00003.6010 1.00003.6025 1.00003.4000
1.00003.1000EA 437 DKK
1.00003.1000 1.00003.2500 1.00003.6010 1.00003.6025 1.00003.4000
Acetonitril, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Acetonitril

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. Merck declare the EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Formel: H₃CCN
Kogepunkt: 81,6 °C (1013 hPa)
Massefylde: 0,782 g/cm³ (20 °C)
Flammepunkt: 2 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001878
CAS-nummer: 75-05-8
EINECS: 200-835-2
UN nummer: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Melting Point -45.7 °C
Molar Mass 41.05 g/mol
Boiling Point 81.6 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point 2 °C
Refractive Index 1.34
Explosion Limit 3.0 - 17 %(V)
Density 0.786 g/cm3 (20 °C)
Ignition Point 524 °C

Specifikationer

Environmentally Preferable Sikrere sundhed for mennesker og miljø Sikrere sundhed for mennesker og miljø
Miljøcertificering Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR