Butyrate standard for IC, TraceCERT®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
08089-100ML
08089-100MLEA 544 DKK
08089-100ML
Butyrate standard for IC, TraceCERT®, Supelco®
Standarder Anioniske standarder Butyrate Standards
Prepared with sodium butyrate and water; filtered through a 0,2 μm membrane filter and stabilised with sodium azide.
Certificeringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.