Polysorbat 20, Reagent Grade

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Sorbitanmonolaurat, ethoxyleret

0777-1LEA 672 DKK
0777-1L 0777-4L
Polysorbat 20, Reagent Grade
Polysorbat 20

Non-ionic detergent for solubilising membrane proteins without altering biological activity.

Formel: C₅₈H₁₁₄O₂₆
Kogepunkt: >100 °C
Massefylde: 1.1 g/cm³ (20 °C)
Flammepunkt: 149 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00165986
CAS-nummer: 9005-64-5
EINECS: 500-018-3

Bestil nu

Specifikationer Testresultater

Arsenic <0.0003%
DNase NONE
Heavy Metals <0.001%
Hydroxyl Number 96 – 108
Identification PASS
Moisture (KF) 3.0%
Protease NONE
Residue on Ignition 0.25%
RNase NONE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR