Multi-element kvalitetskontrolstandard 1 mg/L, 28 elementer i salpetersyre 5% ARISTAR® til ICP
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05200.185EA 1190 DKK
05200.185
Multi-element kvalitetskontrolstandard 1 mg/L, 28 elementer i salpetersyre 5% ARISTAR® til ICP
Standarder Kationiske standarder Multi-Element Standards
28 elements 1 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.
Certificeringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformation: Supplied with certificate of analysis.