Kulstof aktiveret ≥98.5%, powder, Purum ≤40 μm (75%), Fluka™

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Carbon activated, Activated carbon, Activated charcoal

05120-1KG 05120-5KG
05120-1KGEA 1160 DKK
05120-1KG 05120-5KG
Kulstof aktiveret ≥98.5%, powder, Purum ≤40 μm (75%), Fluka™
Kulstof aktiveret

  • ≥95% for general laboratory use.

Formel: C
Smeltepunkt: 3550 °C
Massefylde: 2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00133992
CAS-nummer: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Bestil nu

Specifikationer Testresultater

Loss on drying (105°C) Max. 7 %
Granulation ≤ 40 µm Min.75 % complying
Residue on ignition Max. 7 %
Iron (Fe) Max. 0.3 %
Vapor Pressure <0.1 mmHg ( 20 °C)
Autoignition Temperature 842 °F
Resistivity 1375 μΩ-cm, 20°C (graphite)
Form powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR