Ethidiumbromid ≥98% for biotechnology

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid

0492-25GEA 5150 DKK
0492-25G 0492-5G 0492-CHR-250G 0492-100G
Ethidiumbromid ≥98% for biotechnology
Ethidiumbromid

Fluorescent stain for visualizing nucleic acids.


At a concentration of 0.5 μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10 ng of nucleic acid. At higher concentrations (0.5 to 1.0 μg/ml), it may be used to facilitate purification of DNA in a cesium chloride gradient.


For maximum convenience, VWR offers ethidium bromide in various forms, including ready-to-use solutions and powdered forms. Each is packaged in shatter-resistant packaging for added safety. VWR’s ethidium bromide dropper bottle offers the ultimate convenience and safety. Simply dispense one drop of ethidium bromide into 50ml of agarose gel or running buffer (TBE or TAE) or staining solution.

Formel: C₂₁H₂₀BrN₃
Molekylvægt: 394.31 g/mol
Kogepunkt: 72 °C (0.1 Torr)
Smeltepunkt: 261…264 °C
Massefylde: 1.01 g/cm³ (20 °C)
Flammepunkt: <100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011724
CAS-nummer: 1239-45-8
EINECS: 214-984-6
UN nummer: 2811
ADR: 6.1 I
Merck Index: 13,04751

Bestil nu

Specifikationer Testresultater

DNase None Detected
Loss on Drying ≤5%
Protease None Detected
Purity (Dry Basis) ≥98%
RNase None

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR