BOC-D-Pro-OH

Leverandør: Novabiochem (Part of Merck)
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

8.53095.0025
04-13-0027-25EA 7700 DKK
04-13-0027-25
BOC-D-Pro-OH
BOC-D-Pro-OH
Formel: C₁₀H₁₇NO₄
Molekylvægt: 215.25 g/mol
MDL Number: MFCD00063226
CAS-nummer: 37784-17-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR