Kaliumhydroxid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potash caustic, Caustic potash

0385.1000EA 413 DKK
0385.1000 0385.5000 0385.9050
Kaliumhydroxid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Potassium hydroxide
Formel: KOH
Molekylvægt: 56.11 g/mol
Kogepunkt: 1320
Smeltepunkt: 360
Massefylde: 2.04 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
UN nummer: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Bestil nu

Specifikationer Testresultater

Assay min. 85 %
Aluminium (Al) max. 0.001 %
Calcium (Ca) max. 0.001 %
Sodium (Na) max. 0.5 %
Potassium Carbonate (K2CO3) max. 1.0 %
Silica (SiO2) max. 0.005 %
Trace Impurities (in ppm) Chloride (Cl) max. 5
Trace Impurities (in ppm) Iron (Fe) max. 5
Trace Impurities (in ppm) Nickel (Ni) max. 5
Trace Impurities (in ppm) Heavy Metals (as Pb) max. 5
Trace Impurities (in ppm) Nitrogen Compounds (as N) max. 3
Trace Impurities (in ppm) Phosphate (PO4) max. 5
Trace Impurities (in ppm) Sulfate (SO4) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR