Dodecamolybdophosphorsyre hydrat solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

02553-100ML 02553-500ML
02553-100MLEA 297 DKK
02553-100ML 02553-500ML
Dodecamolybdophosphorsyre hydrat solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®
Dodecamolybdophosphorsyre hydrat

Phosphomolybdic acid solution,ready-to-use spray and plunge reagent, for chromatography (Molybdophosphoric acid)

Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number: MFCD00149913
CAS-nummer: 51429-74-4
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR