Magnesiumchlorid hexahydrat, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Magnesium dichloride

0162.0250EA 299 DKK
0162.0250 0162.1000
Magnesiumchlorid hexahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Magnesiumchlorid hexahydrat
Formel: MgCl₂·6H₂O
Molekylvægt: 203.3 g/mol
Kogepunkt: 1412
Smeltepunkt: 117
Massefylde: 1.57 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149781
CAS-nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Bestil nu

Specifikationer Testresultater

Meets ACS Specifications
Assay 99.0 - 102.0 %
Ammonium (NH4) max. 0.002 %
Barium (Ba) max. 0.005 %
Calcium (Ca) max. 0.01 %
Potassium (K) max. 0.005 %
Sodium (Na) max. 0.005 %
Strontium (Sr) max. 0.005 %
Insoluble Matter max. 0.005 %
Nitrate (NO3) max. 0.001 %
Sulfate (SO4) max. 0.002 %
Trace Impurities (in ppm)
Iron (Fe) max. 5
Manganese (Mn) max. 5
Heavy Metals (as Pb) max. 5
Phosphate (PO4) max. 5

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR