Supelco® Organic Reference Standard, Chlorophyll B

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

00538-1MG
00538-1MGEA 2050 DKK
00538-1MG
Supelco® Organic Reference Standard, Chlorophyll B
Standarder Økologiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf93596cd6a518431696b44e254c1c830e22e368&vid=bf93596cd6a518431696b44e254c1c830e22e368&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 519-62-0
MDL No. MFCD00079053
Formel C₅₅H₇₀MgN₄O₆
Molecular weight 907,47
Flammepunkt Not applicable
Shipping conditions DICE
Storage conditions –20 °C
Opbevaringstemperatur −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR