Chrom ≥99.2% (metals basis), pieces 3-12 mm (0.12-0.47 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

00472.22 00472.36
00472.22EA 1310 DKK
00472.22 00472.36
Chrom ≥99.2% (metals basis), pieces 3-12 mm (0.12-0.47 in)
Chromium

Purity: 99.2% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944

Formel: Cr
Molekylvægt: 52 g/mol
Kogepunkt: 2469
Smeltepunkt: 1840
Massefylde: 7.2 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010944
CAS-nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR