(S)-(+)-2-(Anilinomethyl)pyrrolidine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A0945-1G A0945-5G
TCIAA0945-1GEA 747 DKK
TCIAA0945-1G TCIAA0945-5G
(S)-(+)-2-(Anilinomethyl)pyrrolidine ≥98.0% (by titrimetric analysis)
(S)-(+)-2-(Anilinomethyl)pyrrolidine
Formel: C₁₁H₁₆N₂
Kogepunkt: 126°C
MDL Number: MFCD00015890
CAS-nummer: 64030-44-0
UN nummer: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63c7d9cbd8e554a1ed8e82c361758b474a4e0a64&vid=63c7d9cbd8e554a1ed8e82c361758b474a4e0a64&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +16.0 to +20.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR