4-Brom-2-(trifluormethyl)anilin ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene
A1056-25G
A1056-5G
TCIAA1056-25GEA
1030
DKK
TCIAA1056-25G
TCIAA1056-5G
4-Brom-2-(trifluormethyl)anilin ≥97.0% (by GC)
4-Brom-2-(trifluormethyl)anilin
Formel:
C₇H₅BrF₃N Kogepunkt: 102°C Massefylde: 1,712 g/cm³ (25 °C) Flammepunkt: 107°C |
MDL Number:
MFCD00064393 CAS-nummer: 445-02-3 EINECS: 207-150-8 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...