3,4-Diaminobenzoesyre

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1117-25G D1117-5G
TCIAD1117-25GEA 271 DKK
TCIAD1117-25G TCIAD1117-5G
3,4-Diaminobenzoesyre
3,4-Diaminobenzoesyre
Formel: C₇H₈N₂O₂
Kogepunkt: 354 °C (1013 hPa)
Smeltepunkt: 211 °C
Massefylde: 1,544 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007726
CAS-nummer: 619-05-6
EINECS: 210-577-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5228867adcc1c2a4d393f4801214e157ab6f565e&vid=324b0cf5cb0bd2f43d60affd468c5979ba354c34&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point (Decomposition) 208.0 to 214.0 deg-C
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR