2,6-Dibrom-4-nitroanilin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2414-25G D2414-500G
TCIAD2414-25GEA 353 DKK
TCIAD2414-25G TCIAD2414-500G
2,6-Dibrom-4-nitroanilin ≥98.0% (by GC)
2,6-Dibrom-4-nitroanilin
Formel: C₆H₄Br₂N₂O₂
Smeltepunkt: 204…209 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007639
CAS-nummer: 827-94-1
EINECS: 212-577-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=573d9a3c486bcafa8cb66d01ff38c599d4f9d61a&vid=8e363223bc97a7e3eac493b1c91d858ced3f2fa9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 209.0 deg-C
Solubility in Pyridine almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR