3-Chlor-4-Fluorbenzonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Chlor-3-cyan-6-fluorbenzene

C2081-25G C2081-5G
TCIAC2081-25GEA 1120 DKK
TCIAC2081-25G TCIAC2081-5G
3-Chlor-4-Fluorbenzonitril ≥98.0% (by GC)
3-Chlor-4-Fluorbenzonitril
Formel: C₇H₃ClFN
Kogepunkt: 135°C
Smeltepunkt: 70…71 °C
MDL Number: MFCD00015431
CAS-nummer: 117482-84-5
UN nummer: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcf748c444126798b70cfec9e522bf095048f3a9&vid=dcf748c444126798b70cfec9e522bf095048f3a9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR