2-Fluor-6-iodbenzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
F0660-1G
F0660-5G
TCIAF0660-1GEA
263
DKK
TCIAF0660-1G
TCIAF0660-5G
2-Fluor-6-iodbenzonitril ≥98.0% (by GC)
2-Fluor-6-iodbenzonitril
Formel:
C₇H₃FIN Kogepunkt: 281 °C (1013 hPa) |
MDL Number:
MFCD00015478 CAS-nummer: 79544-29-9 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...