3-(4-Hydroxyphenyl)propionsyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(4-Hydroxyphenyl)propanoic acid, p-hydroxyl phenylpropanol, 3-(p-Hydroxyphenyl)-Propionic Acid
H0599-25G
H0599-250G
TCIAH0599-25GEA
214
DKK
TCIAH0599-25G
TCIAH0599-250G
3-(4-Hydroxyphenyl)propionsyre ≥98.0% (by titrimetric analysis)
3-(4-Hydroxyphenyl)propionsyre
Formel:
C₉H₁₀O₃ Kogepunkt: 208 - 210 °C (14 -rr) Smeltepunkt: 126…129 °C Massefylde: 1,336 g/cm³ (20 °C) |
MDL Number:
MFCD00002778 CAS-nummer: 501-97-3 EINECS: 207-931-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 129.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...