2-(4-Hydroxyphenyl)ethanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

H0720-25G H0720-500G H0720-100G
TCIAH0720-25GEA 279 DKK
TCIAH0720-25G TCIAH0720-500G TCIAH0720-100G
2-(4-Hydroxyphenyl)ethanol ≥98.0% (by GC)
2-(4-Hydroxyphenyl)ethanol
Formel: C₈H₁₀O₂
Kogepunkt: 310°C
Smeltepunkt: 90…92 °C
MDL Number: MFCD00002902
CAS-nummer: 501-94-0
EINECS: 207-930-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e61c90f272d5804f309f426e8423189f851c80b&vid=5e61c90f272d5804f309f426e8423189f851c80b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR