Allyltrimethylsilan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A0729-25ML
A0729-250ML
TCIAA0729-25MLEA
394
DKK
TCIAA0729-25ML
TCIAA0729-250ML
Allyltrimethylsilan ≥98.0% (by GC)
Allyltrimethylsilan
Formel:
H₂CCHCH₂Si(CH₃)₃ Kogepunkt: 84 °C (1013 hPa) Massefylde: 0,7139 g/cm³ (20 °C) Flammepunkt: 7°C |
MDL Number:
MFCD00008635 CAS-nummer: 762-72-1 EINECS: 212-104-5 UN nummer: 1993 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...