2-Brom-p-toluidin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene

B0540-25G B0540-5G
TCIAB0540-25GEA 427 DKK
TCIAB0540-25G TCIAB0540-5G
2-Brom-p-toluidin ≥98.0% (by GC)
2-Brom-p-toluidin
Formel: C₇H₈BrN
Kogepunkt: 240°C
Smeltepunkt: 26 °C
Massefylde: 1,498 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007635
CAS-nummer: 583-68-6
EINECS: 209-515-7
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6094302160b8d84b0583d990a3d4df9ad4bc740&vid=a6094302160b8d84b0583d990a3d4df9ad4bc740&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR