α-Brom-3,4-dichlortoluen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3,4-Dichlorobenzyl bromide

D3700-25G D3700-5G
TCIAD3700-25GEA 1160 DKK
TCIAD3700-25G TCIAD3700-5G
α-Brom-3,4-dichlortoluen ≥98.0% (by GC)
α-Brom-3,4-dichlortoluen
Formel: C₇H₅BrCl₂
Kogepunkt: 86°C
MDL Number: MFCD00041363
CAS-nummer: 18880-04-1
UN nummer: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acecfdc1d776b0e6289e35cb7db7917c2e6d6158&vid=acecfdc1d776b0e6289e35cb7db7917c2e6d6158&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR