α-Brom-3-iodotoluen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-(Bromomethyl)-3-iodobenzene, α-Bromo-m-iodotoulene, 3-Iodobenzyl bromide

I0603-25G I0603-5G
TCIAI0603-25GEA 1820 DKK
TCIAI0603-25G TCIAI0603-5G
α-Brom-3-iodotoluen ≥97.0% (by GC)
α-Brom-3-iodotoluen
Formel: C₇H₆BrI
Smeltepunkt: 50…52 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00019016
CAS-nummer: 49617-83-6
UN nummer: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=951a4367f02e5085e4f771d4e46f55439fe0da42&vid=951a4367f02e5085e4f771d4e46f55439fe0da42&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR