Kapillarkolonner, BPX90
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
054570 054580 054583 054590 054596
558-0347EA 4190 DKK
558-0347 558-0348 558-0351 558-0349 568-0416
Kapillarkolonner, BPX90
Kromatografikolonner
Ideal for fast separation of fragrances, aromatics, petrochemical, pesticides, PCBs and isomers of Fatty Acid Methyl Esters (FAMEs).
90% cyanopropyl polysilphenylene-siloxane
Max. temperature: 80 to 280 °C
Suitable replacement for: Unique to SGE.