TAPS (3-(Tris(hydroxymethyl)methylamino)propan-1-sulfonsyre)
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid
6982.1
6982.2
6982.3
6982.4
ROTH6982.1EA
0
DKK
ROTH6982.1
ROTH6982.2
ROTH6982.3
ROTH6982.4
TAPS (3-(Tris(hydroxymethyl)methylamino)propan-1-sulfonsyre)
TAPS (3-(Tris(hydroxymethyl)methylamino)propan-1-sulfonsyre)
Formel:
C₇H₁₇NO₆S Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007538 CAS-nummer: 29915-38-6 EINECS: 249-954-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...